Assay Details
| Gene Symbol: ptp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026771.3 | NP_001021942.1 | 22 - 23 | 3939 | 123 | |
| NM_001026772.5 | NP_001021943.1 | 10 - 11 | 1892 | 123 | ||
| NM_001026773.5 | NP_001021944.1 | 4 - 5 | 1050 | 123 | ||
| NM_001267341.4 | NP_001254270.1 | 19 - 20 | 3394 | 123 | ||
| NM_001276741.3 | NP_001263670.1 | 23 - 24 | 4011 | 123 | ||
| NM_001276742.3 | NP_001263671.1 | 23 - 24 | 4143 | 123 | ||
| NM_001276743.3 | NP_001263672.1 | 22 - 23 | 4071 | 123 | ||
| NM_001276744.4 | NP_001263673.1 | 4 - 5 | 1698 | 123 | ||
| NM_001276745.3 | NP_001263674.1 | 2 - 3 | 648 | 123 | ||
| NM_001377858.3 | NP_001364824.1 | 23 - 24 | 3681 | 123 | ||
| NM_001377859.2 | NP_001364823.1 | 22 - 23 | 3609 | 123 | ||
| NM_001377860.2 | NP_001364658.1 | 20 - 21 | 3426 | 123 | ||
| NM_001377861.2 | NP_001364506.1 | 20 - 21 | 3134 | 123 | ||
| NM_001377862.2 | NP_001364507.1 | 19 - 20 | 3024 | 123 | ||
More Information
Set Membership: |
|

ptp-3
Genomic Map