Assay Details
| Gene Symbol: ptp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026771.3 | NP_001021942.1 | 23 - 24 | 4219 | 89 | |
| NM_001026772.5 | NP_001021943.1 | 11 - 12 | 2172 | 89 | ||
| NM_001026773.5 | NP_001021944.1 | 5 - 6 | 1330 | 89 | ||
| NM_001267341.4 | NP_001254270.1 | 20 - 21 | 3674 | 89 | ||
| NM_001276741.3 | NP_001263670.1 | 24 - 25 | 4291 | 89 | ||
| NM_001276742.3 | NP_001263671.1 | 24 - 25 | 4423 | 89 | ||
| NM_001276743.3 | NP_001263672.1 | 23 - 24 | 4351 | 89 | ||
| NM_001276744.4 | NP_001263673.1 | 5 - 6 | 1978 | 89 | ||
| NM_001276745.3 | NP_001263674.1 | 3 - 4 | 928 | 89 | ||
| NM_001377858.3 | NP_001364824.1 | 24 - 25 | 3961 | 89 | ||
| NM_001377859.2 | NP_001364823.1 | 23 - 24 | 3889 | 89 | ||
| NM_001377860.2 | NP_001364658.1 | 21 - 22 | 3706 | 89 | ||
| NM_001377861.2 | NP_001364506.1 | 21 - 22 | 3414 | 89 | ||
| NM_001377862.2 | NP_001364507.1 | 20 - 21 | 3304 | 89 | ||
More Information
Set Membership: |
|

ptp-3
Genomic Map