Assay Details
| Gene Symbol: ptp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026771.3 | NP_001021942.1 | 24 - 25 | 4319 | 74 | |
| NM_001026772.5 | NP_001021943.1 | 12 - 13 | 2272 | 74 | ||
| NM_001026773.5 | NP_001021944.1 | 6 - 7 | 1430 | 74 | ||
| NM_001267341.4 | NP_001254270.1 | 21 - 22 | 3774 | 74 | ||
| NM_001276741.3 | NP_001263670.1 | 25 - 26 | 4391 | 74 | ||
| NM_001276742.3 | NP_001263671.1 | 25 - 26 | 4523 | 74 | ||
| NM_001276743.3 | NP_001263672.1 | 24 - 25 | 4451 | 74 | ||
| NM_001276744.4 | NP_001263673.1 | 6 - 7 | 2078 | 74 | ||
| NM_001276745.3 | NP_001263674.1 | 4 - 5 | 1028 | 74 | ||
| NM_001377858.3 | NP_001364824.1 | 25 - 26 | 4061 | 74 | ||
| NM_001377859.2 | NP_001364823.1 | 24 - 25 | 3989 | 74 | ||
| NM_001377860.2 | NP_001364658.1 | 22 - 23 | 3806 | 74 | ||
| NM_001377861.2 | NP_001364506.1 | 22 - 23 | 3514 | 74 | ||
| NM_001377862.2 | NP_001364507.1 | 21 - 22 | 3404 | 74 | ||
More Information
Set Membership: |
|

ptp-3
Genomic Map