Assay Details
| Gene Symbol: ptp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026771.3 | NP_001021942.1 | 25 - 26 | 5937 | 69 | |
| NM_001026772.5 | NP_001021943.1 | 13 - 14 | 3890 | 69 | ||
| NM_001026773.5 | NP_001021944.1 | 7 - 8 | 3048 | 69 | ||
| NM_001267341.4 | NP_001254270.1 | 22 - 23 | 5392 | 69 | ||
| NM_001276741.3 | NP_001263670.1 | 26 - 27 | 6009 | 69 | ||
| NM_001276742.3 | NP_001263671.1 | 26 - 27 | 6141 | 69 | ||
| NM_001276743.3 | NP_001263672.1 | 25 - 26 | 6069 | 69 | ||
| NM_001276744.4 | NP_001263673.1 | 7 - 8 | 3696 | 69 | ||
| NM_001276745.3 | NP_001263674.1 | 5 - 6 | 2646 | 69 | ||
| NM_001377858.3 | NP_001364824.1 | 26 - 27 | 5679 | 69 | ||
| NM_001377859.2 | NP_001364823.1 | 25 - 26 | 5607 | 69 | ||
| NM_001377860.2 | NP_001364658.1 | 23 - 24 | 5424 | 69 | ||
| NM_001377861.2 | NP_001364506.1 | 23 - 24 | 5132 | 69 | ||
| NM_001377862.2 | NP_001364507.1 | 22 - 23 | 5022 | 69 | ||
More Information
Set Membership: |
|

ptp-3
Genomic Map