Assay Details
| Gene Symbol: ptp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026771.3 | NP_001021942.1 | 7 - 8 | 1129 | 86 | |
| NM_001267341.4 | NP_001254270.1 | 4 - 5 | 584 | 86 | ||
| NM_001276743.3 | NP_001263672.1 | 7 - 8 | 1261 | 86 | ||
| NM_001377859.2 | NP_001364823.1 | 7 - 8 | 1129 | 86 | ||
| NM_001377862.2 | NP_001364507.1 | 4 - 5 | 544 | 86 | ||
More Information
Set Membership: |
|

ptp-3
Genomic Map