Assay Details
| Gene Symbol: ptp-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001026771.3 | NP_001021942.1 | 9 - 10 | 1464 | 67 | |
| NM_001267341.4 | NP_001254270.1 | 6 - 7 | 919 | 67 | ||
| NM_001276741.3 | NP_001263670.1 | 10 - 11 | 1536 | 67 | ||
| NM_001276742.3 | NP_001263671.1 | 10 - 11 | 1668 | 67 | ||
| NM_001276743.3 | NP_001263672.1 | 9 - 10 | 1596 | 67 | ||
| NM_001377858.3 | NP_001364824.1 | 10 - 11 | 1536 | 67 | ||
| NM_001377859.2 | NP_001364823.1 | 9 - 10 | 1464 | 67 | ||
| NM_001377860.2 | NP_001364658.1 | 7 - 8 | 951 | 67 | ||
| NM_001377861.2 | NP_001364506.1 | 7 - 8 | 989 | 67 | ||
| NM_001377862.2 | NP_001364507.1 | 6 - 7 | 879 | 67 | ||
More Information
Set Membership: |
|

ptp-3
Genomic Map