Assay Details
| Gene Symbol: csp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027281.1 | NP_001022452.1 | 6 - 7 | 1222 | 60 | |
| NM_001027282.1 | NP_001022453.1 | 2 - 3 | 418 | 60 | ||
| GenBank mRNA | AF088285.1 | - | 8 - 9 | 1394 | 60 | |
| AF088286.1 | - | 2 - 3 | 459 | 60 | ||
More Information
Set Membership: |
|

csp-1
Genomic Map