Assay Details
| Gene Symbol: csp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027281.1 | NP_001022452.1 | 7 - 8 | 1328 | 86 | |
| NM_001027282.1 | NP_001022453.1 | 3 - 4 | 524 | 86 | ||
| GenBank mRNA | AF088285.1 | - | 9 - 10 | 1500 | 86 | |
| AF088286.1 | - | 3 - 4 | 565 | 86 | ||
| AF088287.1 | - | 3 - 4 | 566 | 86 | ||
More Information
Set Membership: |
|

csp-1
Genomic Map