Assay Details
| Gene Symbol: csp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027283.1 | NP_001022454.1 | 2 - 3 | 416 | 61 | |
| GenBank mRNA | AF088287.1 | - | 2 - 3 | 457 | 61 | |
More Information
Set Membership: |
|

csp-1
Genomic Map