Assay Details
| Gene Symbol: pals-23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381682.1 | NP_001367587.1 | 12 - 13 | 1606 | 145 | |
| NM_001384001.1 | 3 - 4 | 527 | 145 | |||
| NM_171857.5 | NP_741063.1 | 4 - 5 | 463 | 145 | ||
| Gene Symbol: pals-24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_064745.5 | NP_497146.2 | 12 - 13 | 1736 | 145 | |
More Information
Set Membership: |
|

pals-23
Genomic Map