Assay Details
| Gene Symbol: pll-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171059.6 | NP_741067.1 | 16 - 17 | 2710 | 70 | |
| NM_171060.5 | NP_741068.1 | 16 - 17 | 2738 | 70 | ||
More Information
Set Membership: |
|

pll-1
Genomic Map