Assay Details
| Gene Symbol: pll-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171059.6 | NP_741067.1 | 19 - 20 | 3094 | 108 | |
| NM_171060.5 | NP_741068.1 | 19 - 20 | 3122 | 108 | ||
More Information
Set Membership: |
|

pll-1
Genomic Map