Assay Details
| Gene Symbol: pll-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171060.5 | NP_741068.1 | 1 - 2 | 115 | 109 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pll-1| Gene Symbol: pll-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171060.5 | NP_741068.1 | 1 - 2 | 115 | 109 | |
Set Membership: |
|