Assay Details
| Gene Symbol: stim-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027745.6 | NP_001022916.1 | 4 - 5 | 650 | 120 | |
| NM_171065.5 | NP_741073.1 | 4 - 5 | 639 | 120 | ||
| NM_171858.5 | NP_741074.1 | 4 - 5 | 639 | 120 | ||
| GenBank mRNA | DQ812088.1 | - | 4 - 5 | 593 | 120 | |
| Gene Symbol: gei-4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001384004.1 | 4 - 5 | 645 | 120 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001384004.1 transcripts.
Set Membership: |
|

stim-1
Genomic Map