Assay Details
| Gene Symbol: dagl-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171860.7 | NP_741085.1 | 12 - 13 | 2783 | 106 | |
| Gene Symbol: T17H7.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381706.2 | NP_001367556.1 | 11 - 12 | 2217 | 106 | |
| Gene Symbol: alh-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381710.1 | NP_001367350.1 | 11 - 12 | 3302 | 106 | |
| NM_001384012.1 | 11 - 12 | 2220 | 106 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381706.2 transcripts.
Set Membership: |
|

dagl-2
Genomic Map