Assay Details
| Gene Symbol: alh-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381710.1 | NP_001367350.1 | 12 - 13 | 3560 | 72 | |
| NM_001384012.1 | 12 - 13 | 2478 | 72 | |||
More Information
Set Membership: |
|

alh-11
Genomic Map