Assay Details
| Gene Symbol: dagl-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027404.4 | NP_001022575.1 | 13 - 14 | 3633 | 76 | |
| NM_171860.7 | NP_741085.1 | 14 - 15 | 3707 | 76 | ||
| Gene Symbol: alh-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381710.1 | NP_001367350.1 | 13 - 14 | 4229 | 76 | |
| NM_001384012.1 | 13 - 14 | 3147 | 76 | |||
More Information
Set Membership: |
|

dagl-2
Genomic Map