Assay Details
| Gene Symbol: dagl-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027404.4 | NP_001022575.1 | 15 - 16 | 3948 | 66 | |
| NM_171860.7 | NP_741085.1 | 16 - 17 | 4022 | 66 | ||
| Gene Symbol: alh-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381710.1 | NP_001367350.1 | 15 - 16 | 4544 | 66 | |
| NM_001384012.1 | 15 - 16 | 3462 | 66 | |||
More Information
Set Membership: |
|

dagl-2
Genomic Map