Assay Details
| Gene Symbol: dagl-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027404.4 | NP_001022575.1 | 6 - 7 | 885 | 91 | |
| NM_001384013.1 | 6 - 7 | 877 | 91 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dagl-2| Gene Symbol: dagl-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027404.4 | NP_001022575.1 | 6 - 7 | 885 | 91 | |
| NM_001384013.1 | 6 - 7 | 877 | 91 | |||
Set Membership: |
|