Assay Details
| Gene Symbol: dagl-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027404.4 | NP_001022575.1 | 8 - 9 | 1617 | 125 | |
| NM_001384013.1 | 8 - 9 | 1609 | 125 | |||
| NM_171860.7 | NP_741085.1 | 9 - 10 | 1688 | 125 | ||
| Gene Symbol: T17H7.7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381706.2 | NP_001367556.1 | 9 - 10 | 1688 | 125 | |
| Gene Symbol: alh-11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001381710.1 | NP_001367350.1 | 9 - 10 | 1687 | 125 | |
| NM_001384012.1 | 9 - 10 | 1691 | 125 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001381706.2, NM_001381710.1, NM_001384012.1 transcripts.
Set Membership: |
|

dagl-2
Genomic Map