Assay Details
| Gene Symbol: zer-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171078.7 | NP_741089.2 | 1 - 2 | 103 | 71 | |
| NM_171079.9 | NP_741090.3 | 1 - 2 | 103 | 71 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

zer-1| Gene Symbol: zer-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171078.7 | NP_741089.2 | 1 - 2 | 103 | 71 | |
| NM_171079.9 | NP_741090.3 | 1 - 2 | 103 | 71 | ||
Set Membership: |
|