Assay Details
| Gene Symbol: zer-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171078.7 | NP_741089.2 | 4 - 5 | 2405 | 70 | |
| NM_171079.9 | NP_741090.3 | 4 - 5 | 2405 | 70 | ||
| Gene Symbol: T24C4.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001384018.1 | 4 - 5 | 2423 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001384018.1 transcripts.
Set Membership: |
|

zer-1
Genomic Map