Assay Details
| Gene Symbol: sax-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171117.5 | NP_741131.2 | 30 - 31 | 8161 | 59 | |
| NM_171118.6 | NP_741132.2 | 5 - 6 | 1645 | 59 | ||
More Information
Set Membership: |
|

sax-2
Genomic Map