Assay Details
| Gene Symbol: cct-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171135.9 | NP_741153.1 | 2 - 3 | 208 | 65 | |
| NM_171136.6 | NP_741154.1 | 2 - 3 | 203 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cct-6| Gene Symbol: cct-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171135.9 | NP_741153.1 | 2 - 3 | 208 | 65 | |
| NM_171136.6 | NP_741154.1 | 2 - 3 | 203 | 65 | ||
Set Membership: |
|