Assay Details
| Gene Symbol: cct-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171136.6 | NP_741154.1 | 5 - 6 | 1226 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cct-6| Gene Symbol: cct-6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171136.6 | NP_741154.1 | 5 - 6 | 1226 | 79 | |
Set Membership: |
|