Assay Details
| Gene Symbol: nmt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_065924.6 | NP_498325.1 | 2 - 3 | 261 | 96 | |
| NM_065925.5 | NP_498326.1 | 2 - 3 | 248 | 96 | ||
| NR_132450.1 | 1 - 2 | 280 | 96 | |||
More Information
Set Membership: |
|

nmt-1
Genomic Map