Assay Details
| Gene Symbol: kin-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027596.4 | NP_001022767.1 | 1 - 2 | 404 | 139 | |
| NM_001379744.1 | NP_001367437.1 | 1 - 2 | 409 | 139 | ||
| GenBank mRNA | U32275.1 | - | 1 - 2 | 412 | 139 | |
More Information
Set Membership: |
|

kin-18
Genomic Map