Assay Details
| Gene Symbol: kin-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027596.4 | NP_001022767.1 | 6 - 7 | 1456 | 67 | |
| NM_001379744.1 | NP_001367437.1 | 6 - 7 | 1458 | 67 | ||
| GenBank mRNA | U32275.1 | - | 6 - 7 | 1464 | 67 | |
More Information
Set Membership: |
|

kin-18
Genomic Map