Assay Details
| Gene Symbol: kin-18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027596.4 | NP_001022767.1 | 7 - 8 | 2198 | 142 | |
| NM_001379744.1 | NP_001367437.1 | 7 - 8 | 2200 | 142 | ||
| GenBank mRNA | U32275.1 | - | 7 - 8 | 2206 | 142 | |
More Information
Set Membership: |
|

kin-18
Genomic Map