Assay Details
| Gene Symbol: akap-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361723.3 | NP_001348743.1 | 11 | 1723 | 64 | |
| NM_001361724.3 | NP_001348742.1 | 10 | 1681 | 64 | ||
| NM_001361725.3 | NP_001348744.1 | 11 - 12 | 1441 | 64 | ||
| NM_001382893.1 | 12 - 13 | 1839 | 64 | |||
| NM_171164.6 | NP_741191.1 | 3 - 4 | 827 | 64 | ||
More Information
Set Membership: |
|

akap-1
Genomic Map