Assay Details
| Gene Symbol: akap-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361723.3 | NP_001348743.1 | 13 | 2058 | 87 | |
| NM_001361724.3 | NP_001348742.1 | 12 | 2016 | 87 | ||
| NM_001361725.3 | NP_001348744.1 | 12 - 13 | 1776 | 87 | ||
| NM_001382893.1 | 13 - 14 | 2174 | 87 | |||
| NM_171164.6 | NP_741191.1 | 4 - 5 | 1162 | 87 | ||
More Information
Set Membership: |
|

akap-1
Genomic Map