Assay Details
| Gene Symbol: akap-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361723.3 | NP_001348743.1 | 1 | 290 | 96 | |
| NM_001361724.3 | NP_001348742.1 | 1 | 290 | 96 | ||
| NM_001382893.1 | 1 - 2 | 388 | 96 | |||
More Information
Set Membership: |
|

akap-1
Genomic Map