Assay Details
| Gene Symbol: akap-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361723.3 | NP_001348743.1 | 5 | 777 | 94 | |
| NM_001361724.3 | NP_001348742.1 | 5 | 777 | 94 | ||
| NM_001361725.3 | NP_001348744.1 | 4 - 5 | 477 | 94 | ||
| NM_001382893.1 | 5 - 6 | 875 | 94 | |||
More Information
Set Membership: |
|

akap-1
Genomic Map