Assay Details
| Gene Symbol: akap-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001361723.3 | NP_001348743.1 | 9 | 1356 | 65 | |
| NM_001361724.3 | NP_001348742.1 | 8 | 1314 | 65 | ||
| NM_001361725.3 | NP_001348744.1 | 8 - 9 | 1074 | 65 | ||
| NM_001382893.1 | 9 - 10 | 1472 | 65 | |||
More Information
Set Membership: |
|

akap-1
Genomic Map