Assay Details
| Gene Symbol: dig-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379776.1 | NP_001366805.1 | 4 - 5 | 402 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

dig-1| Gene Symbol: dig-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001379776.1 | NP_001366805.1 | 4 - 5 | 402 | 71 | |
Set Membership: |
|