Assay Details
| Gene Symbol: ten-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027552.5 | NP_001022723.2 | 9 - 10 | 2216 | 90 | |
| NM_001268011.3 | NP_001254940.1 | 12 - 13 | 2610 | 90 | ||
| NM_001268012.3 | NP_001254941.1 | 10 - 11 | 2388 | 90 | ||
| NM_001379777.2 | NP_001367791.1 | 9 - 10 | 2222 | 90 | ||
| GenBank mRNA | AB206835.1 | - | 9 - 10 | 2216 | 90 | |
| AB206836.1 | - | 4 - 5 | 1722 | 90 | ||
More Information
Set Membership: |
|

ten-1
Genomic Map