Assay Details
| Gene Symbol: ten-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027552.5 | NP_001022723.2 | 12 - 13 | 5074 | 98 | |
| NM_001268011.3 | NP_001254940.1 | 15 - 16 | 5468 | 98 | ||
| NM_001268012.3 | NP_001254941.1 | 13 - 14 | 5246 | 98 | ||
| NM_001379777.2 | NP_001367791.1 | 12 - 13 | 5080 | 98 | ||
| GenBank mRNA | AB206835.1 | - | 12 - 13 | 5074 | 98 | |
| AB206836.1 | - | 7 - 8 | 4580 | 98 | ||
More Information
Set Membership: |
|

ten-1
Genomic Map