Assay Details
| Gene Symbol: prk-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276777.3 | NP_001263706.1 | 3 - 4 | 488 | 100 | |
| NM_001377564.1 | NP_001364690.1 | 2 - 3 | 231 | 100 | ||
| NM_001377565.2 | NP_001364691.1 | 2 - 3 | 231 | 100 | ||
| NM_066127.4 | NP_498528.1 | 2 - 3 | 231 | 100 | ||
More Information
Set Membership: |
|

prk-1
Genomic Map