Assay Details
| Gene Symbol: aco-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066337.7 | NP_498738.2 | 2 - 3 | 178 | 144 | |
| NM_171200.5 | NP_741235.1 | 2 - 3 | 175 | 144 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_066337.7 transcripts.
Set Membership: |
|

aco-2
Genomic Map