Assay Details
| Gene Symbol: aco-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066337.7 | NP_498738.2 | 4 - 5 | 1555 | 78 | |
| NM_171200.5 | NP_741235.1 | 4 - 5 | 1552 | 78 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

aco-2| Gene Symbol: aco-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_066337.7 | NP_498738.2 | 4 - 5 | 1555 | 78 | |
| NM_171200.5 | NP_741235.1 | 4 - 5 | 1552 | 78 | ||
Set Membership: |
|