Assay Details
| Gene Symbol: henn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322688.2 | NP_001309472.1 | 4 - 5 | 996 | 96 | |
| NR_136348.1 | 4 - 5 | 989 | 96 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

henn-1| Gene Symbol: henn-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322688.2 | NP_001309472.1 | 4 - 5 | 996 | 96 | |
| NR_136348.1 | 4 - 5 | 989 | 96 | |||
Set Membership: |
|