Assay Details
| Gene Symbol: pad-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001268141.3 | NP_001255070.1 | 1 - 2 | 378 | 134 | |
| GenBank mRNA | AF455271.1 | - | 1 - 2 | 372 | 134 | |
More Information
Set Membership: |
|

pad-2
Genomic Map