Assay Details
| Gene Symbol: hmt-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001027641.4 | NP_001022812.1 | 4 - 5 | 612 | 117 | |
| GenBank mRNA | AF497513.1 | - | 4 - 5 | 599 | 117 | |
More Information
Set Membership: |
|

hmt-1
Genomic Map