Assay Details
| Gene Symbol: eri-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171244.5 | NP_741292.1 | 4 - 5 | 635 | 64 | |
| NM_171245.5 | NP_741293.1 | 4 - 5 | 635 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

eri-1| Gene Symbol: eri-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171244.5 | NP_741292.1 | 4 - 5 | 635 | 64 | |
| NM_171245.5 | NP_741293.1 | 4 - 5 | 635 | 64 | ||
Set Membership: |
|