Assay Details
| Gene Symbol: eri-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380006.1 | NP_001368344.1 | 2 - 3 | 270 | 121 | |
| NM_001380007.1 | NP_001368343.1 | 2 - 3 | 270 | 121 | ||
| NM_171244.5 | NP_741292.1 | 6 - 7 | 895 | 121 | ||
| NM_171245.5 | NP_741293.1 | 6 - 7 | 895 | 121 | ||
More Information
Set Membership: |
|

eri-1
Genomic Map