Assay Details
| Gene Symbol: eri-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001380006.1 | NP_001368344.1 | 4 - 5 | 639 | 148 | |
| NM_171244.5 | NP_741292.1 | 8 - 9 | 1264 | 148 | ||
More Information
Set Membership: |
|

eri-1
Genomic Map