Assay Details
| Gene Symbol: yars-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171324.5 | NP_741382.1 | 3 - 4 | 349 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

yars-2| Gene Symbol: yars-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_171324.5 | NP_741382.1 | 3 - 4 | 349 | 55 | |
Set Membership: |
|