Assay Details
| Gene Symbol: mgl-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083266.5 | NP_001076735.3 | 13 - 14 | 2446 | 83 | |
| NM_001313057.3 | NP_001299986.1 | 7 - 8 | 1146 | 83 | ||
| NM_001313058.3 | NP_001299987.1 | 7 - 8 | 1146 | 83 | ||
| NM_001313059.3 | NP_001299988.1 | 3 - 4 | 552 | 83 | ||
| NM_001313060.3 | NP_001299989.1 | 3 - 4 | 552 | 83 | ||
| NM_171341.5 | NP_741400.2 | 13 - 14 | 2444 | 83 | ||
| NR_132461.1 | 13 - 14 | 2524 | 83 | |||
| NR_132462.1 | 13 - 14 | 2376 | 83 | |||
| NR_132463.1 | 13 - 14 | 2376 | 83 | |||
More Information
Set Membership: |
|

mgl-3
Genomic Map