Assay Details
| Gene Symbol: mgl-3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083266.5 | NP_001076735.3 | 4 - 5 | 850 | 156 | |
| NM_171341.5 | NP_741400.2 | 4 - 5 | 848 | 156 | ||
| NR_132461.1 | 4 - 5 | 780 | 156 | |||
| NR_132462.1 | 4 - 5 | 780 | 156 | |||
| NR_132463.1 | 4 - 5 | 780 | 156 | |||
More Information
Set Membership: |
|

mgl-3
Genomic Map