Assay Details
| Gene Symbol: drp-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001028204.4 | NP_001023375.1 | 1 - 2 | 314 | 105 | |
| NM_171344.5 | NP_741403.2 | 2 - 3 | 468 | 105 | ||
| GenBank mRNA | AF166274.1 | - | 1 - 2 | 321 | 105 | |
More Information
Set Membership: |
|

drp-1
Genomic Map